Short Path Distillation Kits, United Scientific Supplies
Supplier: United Scientific Supplies
SPDK01 SPD102
76414-748KT 742.52 CAD
76414-748 76414-750
Short Path Distillation Kits, United Scientific Supplies
Distillation Components Distillation Sets
These kits contain essential components for short path distillation.
  • Convenient package
  • High quality 3.3 Borosilicate used for all glass parts
  • Two sizes offered, 250 ml for laboratory and 10000 ml for pilot scale
Short path distillation head with 24/40 flask joints and 10/30 thermometer port (250 ml) features a condenser coil and three ¹/₄" hose nozzles, two for condenser and one for vacuum.
The cow-type distillation receiver has one 24/40 outer joint and three 24/40 inner joints that can be used to attach up to three 50 or 2000 ml single neckMore Product Information