Printed Graduations Beakers, PMP, United Scientific Supplies
Supplier: United Scientific Supplies
P50707 P50708
76414-732CS 379.98 CAD
76414-732 76414-734
Printed Graduations Beakers, PMP, United Scientific Supplies
Beakers
Tapered design is convenient for stacking and holding.
  • Easy to read silk-screened graduations
  • Autoclavable
Polymethylpentene beakers with pouring spouts are ideal for general laboratory use.