Filtering Flasks, United Scientific Supplies
Supplier: United Scientific Supplies
FG5340-500 FG5340-250 FG5340-5000 FG5340-100 FG5340-1000 FG5340-125 FG5340-2000 FG5340-25 FG5340-50
82024-085EA 57.2 CAD
82024-085 82024-087 76414-726 76531-766 76531-768 76531-770 76531-772 76531-774 76531-776
Filtering Flasks, United Scientific Supplies
Flasks Filter Flasks
Heavy wall filtering flask includes a bolt neck with tubulation.
  • Heavy-duty rims
  • Printing in durable white enamel
Borosilicate glass flasks are designed to give the combination of thermal shock resistance and physical strength.