Cotton Tip Applicators, DUKAL™ Corporation
Supplier: Dukal
9003 9006 9013 9016 9026
76407-730BX 18.9 CAD
76407-730 76407-734 76407-736 76407-738 76407-740
Cotton Tip Applicators, DUKAL™ Corporation
Swabs Swabs for Clinical Use
Wood applicators with an absorbent cotton tip. Sterile version is also available with a plastic shaft.
  • Available in non sterile and sterile formats
  • No natural rubber latex
Non sterile applicators are packaged in bags of 100 with a sterilization indicator strip on the bag. Sterile applicators are supplied in easy-peel packages.