SHEL LAB Vacuum Ovens, Sheldon Manufacturing, Inc
Supplier: Sheldon
SLV422 SLV222-EA SLV222 SLV422-EA SLV122-EA SLV122
76414-868EA 15807.66 CAD
76414-868 76414-866 76414-864 76414-872 76414-862 76414-870
SHEL LAB Vacuum Ovens, Sheldon Manufacturing, Inc
Ovens Vacuum Ovens
These SHEL LAB SVAC vacuum ovens provide exceptional capabilities for fast and gentle drying of heat sensitive materials.
  • Stainless steel interiors provide exceptional durability and stability
  • Available in three sizes (from 0.6 to 4.5 cu. ft.)
  • Watlow controller capable of 40 step ramp and soak profiles, or four files with ten steps per file
  • Cross ventilation air flow design
  • Achieve impressive vacuum levels
  • Built-in overtemperature protection
  • 24 month limited warranty
SHEL LAB SVAC vacuum ovens provide exceptional capabilities for fast and gentle drying of heat sensitive materials. They prevent residue accumulation on products and also allow safe drying of materials at lower temperatures than typically required when using convection ovens. All SHEL LAB vacuum ovens are built with a stainless steel chamber for exceptional durability. Our double plenum design meets UL, CSA and EU safety requirements resulting in a cool outer surface. The cross-flow ventilation forces inert gas to fill the entire chamber. To achieve required vacuum levels, users can choose from a ³/₈" orifice or a KF-25 fitting to withstand heavy use and minimize draw-down time.
The doors on these units have positive latch handles with spring-loaded glass to facilitate a good vacuum seal without hinge binds that shorten the gasket life. The tempered glass window allows for safe, continuous monitoring of samples.
The SHEL LAB Vacuum Ovens are equipped with silicone gaskets. While these gaskets resist a wide variety of temperatures and pressures, they can breakdown in the presence of solvents and acids. For solvent applications, we recommend using Buna gaskets. For acidic applications, we recommend using Fluorosilicone or Viton® gaskets.
Certifications: Meets UL, CSA, and EU safety requirements.
Ordering information: Does not include pump. Please contact Avantor for assistance with choosing the ideal pump.