Rat Recombinant IL-2 (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
300-38AF-20UG 300-38AF-100UG 300-38AF-500UG 300-38AF-1MG
75845-240EA 368.28 CAD
75845-240 75845-242 75845-244 75845-246 75845-306
Rat Recombinant IL-2 (Animal free) (from E. coli)
Proteins and Peptides
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes.  IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
  • High Quality
  • Low Endotoxin
  • Affordable
Caution: For research use only.