Muffle Furnaces up to 1400 °C, Nabertherm
Supplier: NABERTHERM INC.
L-054U4LN10-240 L-054U4ON10-240 L-054V4LN10-240 L-054V4ON10-240 L-094U4LN8-208 L-094U4LN8-240 L-094U4ON8-208 L-094U4ON8-240 L-094V4LN8-208 L-094V4LN8-240 L-094V4ON8-208 L-094V4ON8-240 L-154U4LN11-240 L-154U4ON11-240 L-154V4LN11-240 L-154V4ON11-240
76537-784EA 12286.76 CAD
76537-784 76537-786 76537-788 76537-790 76537-792 76537-794 76537-796 76537-798 76537-800 76537-802 76537-804 76537-806 76537-808 76537-810 76537-812 76537-814 76537-816 76537-818 76537-820 76537-822 76537-824 76537-826 76537-828 76537-830
Muffle Furnaces up to 1400 °C, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These Muffle furnaces stand out for their excellent workmanship, advanced and attractive design, and high level of reliability. Heating elements on support tubes radiating freely into the furnace chamber provide particularly short heating times and a maximum temperature of 1400 °C. These models are a good alternative to the familiar L(T) ../11 series when higher application temperatures are needed.
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Dual shell housing for low external temperatures and
More Product Information