Sterile ABD Pads, DUKAL™ Corporation
Supplier: Dukal
5590 5810
76407-668BX 21.79 CAD
76407-668 76407-670 76407-672
Sterile ABD Pads, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Soft non-woven outer facing wicks moisture into a cellulose core, which quickly absorbs and disperses fluid.
  • Highly absorbent multi-layer dressing
  • Moisture resistant barrier prevents fluid strike-through
  • No natural rubber latex
All edges are sealed to prevent linting and leakage.