Conforming Stretch Gauze, DUKAL™ Corporation
Supplier: Dukal
601 602 603 604 606 701 702 703 704 706
76407-574BG 5.45 CAD
76407-574 76407-576 76407-578 76407-580 76407-582 76407-590 76407-592 76407-594 76407-596 76407-598
Conforming Stretch Gauze, DUKAL™ Corporation
First Aid Supplies Wound Dressings
Rayon/poly knitted stretch gauze bandage.
  • Specifically designed not to slip while in use
  • Available in non sterile and sterile formats
  • No natural rubber latex
Conforming stretch gauze provides slight compression to the site while securely holding to any body contour. The unique weave construction and finished edges prevent unraveling and linting.
Order Now
SPECIFICATIONS
Environmentally Preferable