SCOUT® SJX/E Class II Legal-for-Trade Portable Balances, OHAUS

Supplier: Ohaus
30253056 30253055
76414-148EA 2474.88 CAD
76414-148 76414-150
SCOUT® SJX/E Class II Legal-for-Trade Portable Balances, OHAUS
Balances and Scales
Designed for a wide range of Legal-for-Trade applications, the Class ll and Class III certified SJX balances are ideal for weighing cannabis, jewellery, precious metals and gemstones.

  • Class ll Legal-for-Trade compliant balances suitable for countertop retail applications
  • Manual internal settings allow easy calibration
  • Advanced weighing technology gives reliable, repeatable weighing results
  • Application modes are weighing, parts counting, percent weighing, check weighing, totalization, density determination, display hold
  • Weighing units are g, kg, ct, oz., ozt, dwt, lb, grn

Legal-for-Trade approvals: NTEP and Measurement Canada approved models meet or exceed Class II (SJX1502N/E) or III (SJX6201N/E) accuracy requirements, making the Scout ideal for commercial applications where reliability and precision are essential.

The superior mechanical overload protection system of the Scout is comprised of a high-strength stainless steel pan and an impact-resistant plastic pan support. This enables the balance to safely withstand loads up to 10 times its rated capacity.

The large backlit LCD gives operators a clear view of their weighing results. An optional auxiliary display can also be installed easily to provide additional weight notifications to customers in retail environments.

Stabilization time as fast as 1 second and high resolution load cell with up to 150000 d resolution and readability to 0.001 g, deliver extremely quick, precise and repeatable weighing results.

Scout has multiple connectivity options for convenient data logging and communication, as well as connecting to an auxiliary display. Options include - RS232, USB hlost, USB device, ethernet and Bluetooth®. With the USB host option, the user can simply save and transfer data from the balance to a USB drive. The bluetooth option allows you to wirelessly operate the balance via a mobile device or tablet for Android™.

Certifications: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, IEC/EN 61326-1 Class B; FCC Part 15 Class B; Industry Canada ICES-003 Class B.

Ordering information: Accessories not included.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR