HelixMark® Special Silicone Tubing, Dow Corning Silicone Material, Freudenberg Medical
Supplier: Freudenberg Medical
61-759-82 61-763-60
CA11008-496EA 4150.5 CAD
CA11008-496 CA11008-498
HelixMark® Special Silicone Tubing, Dow Corning Silicone Material, Freudenberg Medical
Tubing Transfer Tubing
HelixMark special silicone tubing, Dow Corning silicone material longer length.
  • Certified manufacturing in compliance with ISO 13485 quality standards
  • Sterilizable in autoclaves, with ethylene oxide and by gamma irradiation
  • Material certification in every package
  • Lot traceability
  • Double bagged and box packaging
  • 50 Shore A durometer
Tubing is manufactured in Freudenberg Medical's FDA registered medical device facility following Good Manufacturing Practies (GMPs) in accordance with Quality Systems Regulation.The manufacturing process minimizes flow degradation over extended periods.
Certifications: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177-2600 and EP 3.1.9 or EP 3.1.9 without appearance of solution. Meets USP Class VI biocompatibility requirements.
Accessories information: Accessories not included.
Ordering information: Supplied in spool.
Packaging: Double bagged, individual box packaging.