Kjeldahl Weighing Boats, VELP Scientifica
Supplier: VELP SCIENTIFIC INC.
CM0486000 CM0486001
10184-066EA 616.36 CAD
10184-066 10184-068
Kjeldahl Weighing Boats, VELP Scientifica
Boats Weighing Boats
Weighing boat is suitable for the syrupy and viscose substances, pulps and powders.
  • Nitrogen-free boat, ensures low error risk
  • Suitable for quantitative transfer of substance into mineralization test tubes