Nitazoxanide ≥98% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide, 2-((5-Nitrothiazol-2-yl)carbamoyl)phenyl acetate, NTZ

N1031-1G N1031-200MG N1031-5G
TCN1031-1GEA 382.36 CAD
TCN1031-1G TCN1031-200MG TCN1031-5G
Nitazoxanide ≥98% (by HPLC, titration analysis)
Nitazoxanide
Formula: C₁₂H₉N₃O₅S
Storage Temperature: Refrigerator
MDL Number: MFCD00416599
CAS Number: 55981-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f86a77241736b2597290c617efa1448f48ff850&vid=3f86a77241736b2597290c617efa1448f48ff850&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity(HPLC) min. 98.0 area%
Purity(Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR