Nelfinavir mesylate hydrate ≥97% (by HPLC)

Supplier: TCI America

Synonyms: AG-1343, Nelfinavir methanesulphonate hydrate, Nelfinavir methanesulfonate hydrate

N0986-100MG N0986-25MG
TCN0986-100MGEA 657.28 CAD
TCN0986-100MG TCN0986-25MG
Nelfinavir mesylate hydrate ≥97% (by HPLC)
Nelfinavir mesylate hydrate
Formula: C₃₂H₄₅N₃O₄S·CH₄O₃S·xH₂O
MW: 663.89 g/mol (anhy)
MDL Number: MFCD00931436
CAS Number: 159989-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc05f7d807ef3a8cecf42ab65ffa1a095a765140&vid=1ab401671b7442f7c926bb47a5fd7e4eae59c757&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity(HPLC) min. 97.0 area%
Specific rotation [a]20/D -109 to -113 deg(C=1, MeOH)(calcd.on anh.substance)
Water 2.0 to 8.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR