Inosine Pranobex ≥98% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Acetamidobenzoic acid,9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one,1-(Dimethylamino)propan-2-ol (3:1:3)

I1037-100MG I1037-1G
TCI1037-100MGEA 42.66 CAD
TCI1037-100MG TCI1037-1G
Inosine Pranobex ≥98% (by HPLC, titration analysis)
Inosine Pranobex
Formula: C₁₀H₁₂N₄O₅·₃C₉H₉NO₃·3C₅H₁₃NO
MW: 1115.25 g/mol
MDL Number: MFCD05662374
CAS Number: 36703-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2afa6f0030be60d6b33aedb48286b3da604bf90&vid=b2afa6f0030be60d6b33aedb48286b3da604bf90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity(HPLC) min. 98.0 area%
Purity(Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -9.0 to -13.0 deg(C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR