3-Hydroxypyrazine-2-carboxamide ≥98% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: T-1105

H1485-1G H1485-5G
TCH1485-1GEA 60.04 CAD
TCH1485-1G TCH1485-5G
3-Hydroxypyrazine-2-carboxamide ≥98% (by HPLC, titration analysis)
3-Hydroxypyrazine-2-carboxamide
Formula: C₅H₅N₃O₂
MW: 139.114 g/mol
MDL Number: MFCD00233977
CAS Number: 55321-99-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=712c9d4386eeabe05c2f043eb27b1592461f0860&vid=712c9d4386eeabe05c2f043eb27b1592461f0860&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity(HPLC) min. 98.0 area%
Purity(Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR