(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine ≥98% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: PSI-6206, 1-[(2R,3R,4R,5R)-3-Fluoro-4-hydroxy-5-(hydroxymethyl)-3-methyloxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

D4823-1G D4823-200MG
TCD4823-1GEA 636.42 CAD
TCD4823-1G TCD4823-200MG
(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine ≥98% (by HPLC, titration analysis)
(2'R)-2'-Deoxy-2'-fluoro-2'-methyluridine
Formula: C₁₀H₁₃FN₂O₅
MW: 260.219 g/mol
Storage Temperature: Freezer
MDL Number: MFCD27918669
CAS Number: 863329-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25c84dc4bb208388b1195e4f098f949c8072139b&vid=566cbd77304de3714f1b7c84e93b6a482137e542&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity(HPLC) min. 98.0 area%
Purity(Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +87.0 to +92.0 deg(C=1,MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR