Nalgene® Validation Bottles, PETG, Clean, Certified, Thermo Scientific
Supplier: Thermo Fisher Scientific
382035-0005 382035-0020
76397-858CS 2900.99 CAD
76397-858 76411-626
Nalgene® Validation Bottles, PETG, Clean, Certified, Thermo Scientific
Bottles Plastic Bottles
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • PETG construction
These bottles can reduce the risk of contamination.
Certifications: Validated to USP 788 and <0.5 EU/ml endotoxin.
Order Now
SPECIFICATIONS
Environmentally Preferable