LABeler™ and LinkLabel™ Label Printers
Supplier: MTC BIO, INC.
LABeler™
L3000 L9010-24WK L9010-6CK L9010-12WK L9010-24ULT L9010 L9010-6WK L9010-12CK L9010-12GW L9010-18WK L9010-12BW L9010-PS L9010-12ULT L9010-12YK L9010-24CK L9010-12RW
470325-960EA 88.6 CAD
470325-960 470325-946 470325-956 470325-934 470325-944 470325-954 470325-932 470325-942 470325-930 470325-952 470325-940 470325-950 470325-938 470325-948 470325-958 470325-936
LABeler™ and LinkLabel™ Label Printers
Label Makers
Select from two high quality labelers and a wide range of label options.
  • Print barcodes, QR codes, text, or serial numbers
  • Adjustable label lengths minimize waste
  • Hardy labels hold up under the harshest conditions
The LABeler™ Lab printer provides the convenience of a lab marking pen with the benefits of a large desktop printer. Simply type in the desired information using the 50+ button key pad and press the print key. Press the cut level and the label is instantly ready to apply. Use the included power supply or for extra portability, the unit can run on 6 AA batteries (not included).
The LinkLabel™ Lab printer can be easily programmed from a phone via Bluetooth connection. Just download the free LabelPro™ app to endlessly customize and specialize printed labels. It comes pre-loaded with hundreds of scientific symbols, and can print barcodes, QR codes, text, or numbers. Use the included 115/230V Universal power supply or for extra portability, the unit can run on AA batteries (not included). LinkLabel™ comes with one white label cartridge.
A wide-range of labels are compatible with either printer and come in several different widths. The 6 mm tapes are great for tissue culture plates, 12 mm tapes are ideal from microtubes, and the extra tall 24 mm tapes are ideal for large tubes and flasks. Standard label tapes can be autoclaved, boiled, or even frozen to temperatures as low as −50 °C. For more durability, ULT label tapes are great for use with liquid nitrogen and can withstand environments as cold as −196 °C.
Certifications: cUL listed.