VWR® E-Series Balances with Calibration Certificate
Supplier: VWR International
76378-350 76378-436 76378-452 76378-454 76378-456 76378-458
76378-350EA 2914.54 CAD
76378-350 76378-436 76378-452 76378-454 76378-456 76378-458
VWR® E-Series Balances with Calibration Certificate
Balances and Scales
The VWR® E-Series/C balances with calibration certificate are the answer for an organizations regulatory and compliance needs.
  • Complete calibration data reported
  • Backlit, LCD screen
  • External calibration
  • RS232 communication interface
  • Five year warranty
The portable balances include a Certificate of Calibration showing the full testing data from an ISO/IEC 17025:2017 accredited Calibration and Testing Laboratory utilizing NIST-traceable weights. These economical and versatile balances offer a variety of standard features, such as multiple weighing units,More Product Information
Order Now
SPECIFICATIONS
Operating temperature range 0…40 °C (32…104 °F)
Environmentally Preferable
Communication interface RS232
Voltage 100 - 240 VAC