PIPETMAN® Multichannel Mechanical Pipettor, Gilson
Supplier: GILSON, INC.
F144068 F144069 F144070 F144071 F144072 F144073 F144074 F144075
76408-356EA 1565.11 CAD
76408-356 76408-358 76408-360 76408-362 76408-364 76408-366 76408-368 76408-370
PIPETMAN® Multichannel Mechanical Pipettor, Gilson
Pipettes Micropipettes
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling. PIPETMAN pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting.
  • Legendary performance
  • Easy pipetting
  • Built-to-last and convenient
  • Simple to maintain
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the worldMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable