Methanol, anhydrous 99.9%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger


Packaged under argon.

047339-AK 047339-K2
AA47339-AKEA 83.33 CAD
AA47339-AK AA47339-K2
Methanol, anhydrous 99.9%, AcroSeal™
Methanol

Packaged under argon in resealable AcroSeal® bottles.

Formula: CH₃OH
Boiling Pt: 64.7 °C
Melting Pt: –98 °C
Density: 0.791
Flash Pt: 11 °C (52 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
Merck Index: 14,5957
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR