Quick-DNA/RNA Viral MagBead, Zymo Research
Supplier: Zymo Research
Quick-DNA/RNA Viral MagBead kit is designed for high-throughput purification of viral DNA and/or RNA from plasma, serum, urine, cell culture media, blood, saliva, cellular suspensions, biopsies and swab and fecal samples stored in DNA/RNA Shield (for sample collection, nucleic acid preservation and inactivation of pathogens).
- High-throughput, magnetic-bead based purification of viral DNA and RNA from plasma, serum, urine, cell culture media, blood, saliva, cellular suspensions, swab, fecal and biopsy samples
- High-quality DNA/RNA ready for Next-Gen sequencing, RT/qPCR, hybridization, etc
- DNA/RNA Shield included for sample collection, inactivation and storage
The kit features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and eluted.
Isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.
Materials: ZR-96 MagStand, Collection Plate, 96-Well Block, Elution Plate, Cover Foil
Certifications: Meets ASTM standards. EN 342. Quick-DNA/RNA Viral MagBead – DX are CE, IVD certified.
Accessories information: ZR-96 MagStand (VWR# 76005-120)
Collection Plate (VWR# 76005-214; capacity 1.2 ml/well)
96-Well Block (VWR#s 76005-116, 76004-722, 76005-118)
Elution Plate (VWR# 76004-222; capacity 0.35 ml/well)
Cover Foil ( VWR#s 76004-236, 76005-218, 76005-220, 76005-222, 76004-234, 76005-216)
Ordering information: Equipment needed: Magnetic stand or separator, liquid handler or robotic sample processor (user provided).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...