Performance Plastic Chemical Wash Bottles, 500 ml, 1 Color Label, Eisco Scientific
Supplier: Wards
CHWB1010 CHWB1019 CHWB1028 CHWB1037 CHWB1046 CHWB1010PK2 CHWB1019PK2 CHWB1028PK2 CHWB1037PK2 CHWB1046PK2 CHWB1010PK6 CHWB1019PK6 CHWB1028PK6 CHWB1037PK6 CHWB1046PK6
470323-920EA 8.05 CAD
470323-920 470323-932 470323-944 470323-956 470323-968 470323-986 470323-998 470324-010 470324-022 470324-034 470324-052 470324-064 470324-076 470324-088 470324-100
Performance Plastic Chemical Wash Bottles, 500 ml, 1 Color Label, Eisco Scientific
Bottles Wash Bottles
Innovative performance plastic and flex ink for durable, easy to use, chemical specific laboratory wash bottles.
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
Wash bottles are made with performance plastic, a low density polyethylene (LDPE) for superior quality, strength, andMore Product Information