Performance Plastic Wash Bottles, No Label, Eisco Scientific
Supplier: Wards
CHWB1001 CHWB1002 CHWB1003 CHWB1001PK2 CHWB1002PK2 CHWB1003PK2 CHWB1001PK6 CHWB1002PK6 CHWB1003PK6
470323-914EA 7.05 CAD
470323-914 470323-916 470323-918 470323-980 470323-982 470323-984 470324-046 470324-048 470324-050
Performance Plastic Wash Bottles, No Label, Eisco Scientific
Bottles Wash Bottles
Innovative performance plastic and flex ink for a durable and easy to use laboratory wash bottles.
  • Unlabeled chemical wash bottle
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Self- venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
  • Versatile staple for every laboratory
Wash bottles are made with Eisco Labs exclusive Performance Plastic, a low density polyethylene (LDPE) designed for superior quality and strength. No label design provides a space to write contents. Each bottle features a two-piece construction, consisting of the bottle and leak-proof cap with built-in nozzle. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
Packaging: Bottles are available individually and in packs of two or six.