H-Flex 1 Swinging Bucket Rotor, Thermo Scientific
Supplier: Thermo Fisher Scientific
75003300 75003301
76404-696EA 4250.2 CAD
76404-696 76404-698
H-Flex 1 Swinging Bucket Rotor, Thermo Scientific
Centrifuges Rotors
Increased flexibility in the Thermo Scientific ™ general purpose 1.6 L benchtop centrifuges.
  • Autolock and click seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Spin more with less. Whether clinical and diagnostic applications or cell culture and other research,users can now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint general purpose centrifuge. High capacity and flexibility - all with a small footprint. Compatible with the megafuge 16 and multifuge X1 general purpose centrifuge platforms.