Performance Plastic Chemical Wash Bottles, 1000 ml, 2 Color Label, Eisco Scientific
Supplier: Wards
CHWB1014 CHWB1023 CHWB1032 CHWB1041 CHWB1050 CHWB1014PK2 CHWB1023PK2 CHWB1032PK2 CHWB1041PK2 CHWB1050PK2 CHWB1014PK6 CHWB1023PK6 CHWB1032PK6 CHWB1041PK6 CHWB1050PK6 CHWB1059
470323-928EA 16.6 CAD
470323-928 470323-940 470323-952 470323-964 470323-976 470323-994 470324-006 470324-018 470324-030 470324-042 470324-060 470324-072 470324-084 470324-096 470324-108 470324-116
Performance Plastic Chemical Wash Bottles, 1000 ml, 2 Color Label, Eisco Scientific
Bottles Wash Bottles
Innovative performance plastic and flex ink for durable, easy to use, chemical specific laboratory wash bottles.
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
Wash bottles are made with performance plastic, a low density polyethylene (LDPE) for superior quality, strength, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable