VWR® Irradiated Vinyl Cleanroom Tapes
Supplier: VWR International
76311-314 76311-316 76311-318 76311-320
76311-314CS 1275.19 CAD
76311-314 76311-316 76311-318 76311-320
VWR® Irradiated Vinyl Cleanroom Tapes
Tapes
An irradiated vinyl tape with low particulate and bio-burden counts for use in critical and aseptic cleanroom applications, including sealing environmental monitoring plates and clean construction areas.
  • Low particulate and bio-burden counts
  • Ideal for sealing, color-coding, and marking in aseptic area
  • Irradiated, pressure-sensitive vinyl tape
  • Processed in a cleanroom
  • Individually double bagged
  • Can be written on with cleanroom pen
  • Available in a range of widths
  • 3" plastic core
  • Roll length: 33 m (36 yard)
Processed in a controlled environment, wound on a plastic core, and individually double bagged. Each roll is 36 yards and available in 0.5, 0.75, 1, or 2" widths.
Delivery information: Supplied with full certification of irradiation and lot traceability.