Performance Plastic Chemical Wash Bottles, 1000 ml, 4 Color Label
Supplier: Wards
CHWB1015 CHWB1024 CHWB1033 CHWB1042 CHWB1051 CHWB1015PK2 CHWB1024PK2 CHWB1033PK2 CHWB1042PK2 CHWB1051PK2 CHWB1015PK6 CHWB1024PK6 CHWB1033PK6 CHWB1042PK6 CHWB1051PK6 CHWB1060
470323-930EA 18.15 CAD
470323-930 470323-942 470323-954 470323-966 470323-978 470323-996 470324-008 470324-020 470324-032 470324-044 470324-062 470324-074 470324-086 470324-098 470324-110 470324-118
Performance Plastic Chemical Wash Bottles, 1000 ml, 4 Color Label
Bottles Wash Bottles
Innovative performance plastic and flex ink for durable, easy to use, chemical specific laboratory wash bottles.
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
Wash bottles are made with performance plastic, a low density polyethylene (LDPE) for superior quality, strength, and flexibility. Each bottle is color coded and labeled for a specific chemical featuring that chemical's information as well as hazard, safety and first aid precautionary statements. Color coded leak-proof caps with built-in nozzle included. The low density polyethylene plastic is very responsive, with smooth bounce back that won't get stiff over time. Easy to control squeeze for dispensing.
Sodium Hypochlorite (bleach) wash bottles are opaque (white) to protect contents from UV light rays. Other bottles are clear/transparent.
Certifications: Bottle labeling complies with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS), OSHA’s HazCom 2012 requirements, and OSHA requirements as outlined in Hazard Communication Standard 29 CFR 1910.1200.
Packaging: Bottles are available individually and in packs of two or six.