Performance Plastic Chemical Wash Bottles, 500 ml, 4 Color Label
Supplier: Wards
CHWB1012 CHWB1021 CHWB1030 CHWB1039 CHWB1048 CHWB1012PK2 CHWB1021PK2 CHWB1030PK2 CHWB1039PK2 CHWB1048PK2 CHWB1012PK6 CHWB1021PK6 CHWB1030PK6 CHWB1039PK6 CHWB1048PK6 CHWB1057
470323-924EA 12.8 CAD
470323-924 470323-936 470323-948 470323-960 470323-972 470323-990 470324-002 470324-014 470324-026 470324-038 470324-056 470324-068 470324-080 470324-092 470324-104 470324-114
Performance Plastic Chemical Wash Bottles, 500 ml, 4 Color Label
Bottles Wash Bottles
Innovative performance plastic and flex ink for durable, easy to use, chemical specific laboratory wash bottles.
  • Eisco exclusive Performance Plastic is durable yet extremely responsive
  • Made of low-density polyethylene (LDPE)
  • Easy to read labeling, including chemical specific information and safety warnings
  • Self-venting design to eliminate leaks and dripping
  • Wide mouth for easy filling
  • Innovative flex ink won’t fade, crack, or peel
Wash bottles are made with performance plastic, a low density polyethylene (LDPE) for superior quality, strength, andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable