VWR® Wire Racks
Supplier: VWR International
76358-652 76358-654 76358-656 76358-658 76358-660 76358-662 76358-664 76358-666 76358-668 76358-670 76358-672 76358-674 76358-676 76358-678 76358-680
76358-652CS 1345.53 CAD
76358-652 76358-654 76358-656 76358-658 76358-660 76358-662 76358-664 76358-666 76358-668 76358-670 76358-672 76358-674 76358-676 76358-678 76358-680
VWR® Wire Racks
Racks Tube Racks
These HDPE coated wire racks are durable for transport or sample handling.
  • Chemically-resistant
  • Racks can be used between temperatures of –40 °C and 121 °C
  • Slotted bottoms
  • Holds all brands and types of tubes/bottles for each size range
  • Square wells and three deck levels help keep tubes secure
  • Autoclavable
Suitable for conical, round or flat-bottom glass or plastic tubes and bottles.
Order Now
SPECIFICATIONS
Environmentally Preferable