Shark Week Kits
Supplier: GENOSENSOR CORPORATION
7003 7004
470325-172EA 294.2 CAD
470325-172 470325-174
Shark Week Kits
Educational Classroom Kits and Activities
Teach fundamental concepts of genetic diversity and real world applications in marine biology.
Marine biologist have identified a new species of shark that washed up on the beach. In an attempt to learn as much about the shark as they can, they have decided to analyze the sharks stomach contents to identify its main food source.
Plasmid DNA is used to emulate the genetic diversity seen in biology. Four plasmid samples are included in the kit, one representing DNA from the main food source, and three for comparison against the unknown food DNA.
Kit I: Allows students to perform aMore Product Information