EduPanel LED Lights
Supplier: SI Manufacturing
90152 90102
470231-748EA 345.05 CAD
470231-748 470231-750
EduPanel LED Lights
Transilluminators LED Transilluminators
Featuring an ultra slim design and dimmable LED technology.
  • LED technology
  • Ultra-slim and lightweight
  • Two sizes available
EduPanel features a 50000 hour LED life span, and provides a bright, illuminated background that's perfect for lighting items from beneath. EduPanel works well with transparent and translucent materials like X-rays and electrophoresis gels.
This portable panel adds excitement and illumination to various STEM related topics including patterning, shapes, geometry, color exploration, optics, opacity, transparency, tracing, stenciling and more. Dimensions listed represent viewing area size.
Delivery information: Includes AC power adapter.