THGA Furnace Tube Inspection Mirror, PerkinElmer
Supplier: PerkinElmer
B0851900 B0851900
CA97013-696EA 223.34 CAD
97013-696 CA97013-696
THGA Furnace Tube Inspection Mirror, PerkinElmer
Mirrors
This mirror is designed to observe the sample during an THGA furnace temperature programmed run. By observing the sample during the run, the drying temperatures and times can be optimized.
Ordering information: HGA instrument mirrors are available separately.