Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Supplier: PerkinElmer
N1011446 N1011446
CA97019-184EA 117.4 CAD
CA97019-184 97019-184
Zero Dilution Inner Liner, 2 ext.Ø.×73L mm, PerkinElmer
Chromatography Supplies Chromatography Inlet Liners
To be used in conjunction with outer liner (N1011447).
  • Not compatible with the Clarus 590/690 Capillary Injectors
  • 73 mm length
Inlet liners for split injection have mixing chambers with tortuous flow paths to allow full vaporization of the sample. Deactivating the surface of these liners prevents active compounds from degrading. Packing the liner with wool will trap non-volatile residue and prevent column contamination when analyzing dirty samples.