Worktables, SPEC-LINE Series with Flat Top, Undershelf Style, Advance Tabco
Supplier: Advance Tabco
VSS-305 VLG-304 VLG-306
58300-022EA 10183.34 CAD
58300-022 58300-026 58300-030
Worktables, SPEC-LINE Series with Flat Top, Undershelf Style, Advance Tabco
Furniture Tables and Workbenches
Top is furnished with a 2×1" square die embossed NO-DRIP countertop edge with a ¹/₂" return on four sides.
  • 14 gauge stainless steel type '304' series top
  • Galvanized or stainless steel legs and 18 gauge undershelf
  • NSF
To reinforce and maintain a level working surface, 24" wide tables are supplied with two hat channels and 30" and 36" wide tables are supplied with three hat channels. Pre-engineered welded angle adapters insure ease of future drawer installation.
Aluminum die cast 'leg-to-shelf' clamp secures shelf to leg eliminating unsightly nuts and bolts. Undershelf is adjustable.