Worktables, Stainless Steel, Hinged Door Style with Enclosed Base, Advance Tobaco
Supplier: Advance Tabco
HB-SS-306 HB-SS-305
58300-060EA 22937.89 CAD
58300-060 47744-678
Worktables, Stainless Steel, Hinged Door Style with Enclosed Base, Advance Tobaco
Furniture Tables and Workbenches
Flat top option is furnished with a 1⁵/₈" sanitary rolled rim on the front and 1⁵/₈" square bend edges on the two sides and rear.
  • Enclosed cabinet base with hinged doors
  • 14 gauge Type 304 stainless steel top
  • Bolt-on stainless steel leg
  • NSF
Ordering information: Enclosed base modifications and accessories are available; contact your Avantor representative for more information.