Square Corner Scullery, Stainless Steel, Advance Tobaco
Supplier: Advance Tabco
4-81-18 4-2-48
47744-686EA 3519.2 CAD
47744-686 47744-690
Square Corner Scullery, Stainless Steel, Advance Tobaco
Faucets and Sinks Sinks
The '400' series is 16 gauge type 430 series.
  • All heli-arc welded
  • Galvanized tubular legs with 1" plastic bullet feet
Welded areas blended to match adjacent surfaces and to a satin finish. Supply is ¹/₂" hot and cold. Faucet holes on 8" O.C. Waste drains are 1¹/₂" IPS basket type and are included.
Ordering information: Faucets are not included.