DMiL Inverted Microscope Packages, Leica Microsystems
Supplier: Leica Microsystems
11541544 11521288 11504254 11504127 11506408 11506272 11506298 11506203 11504190 11504174 11504179 11504208 11504117
CA10037-632EA 809.75 CAD
CA10037-632 CA76377-688 CA76377-690 CA76377-696 CA76377-698 CA76377-700 CA76377-702 CA76377-704 CA76377-706 CA76377-708 CA76377-710 CA76377-712 CA76377-714 CA10037-662
DMiL Inverted Microscope Packages, Leica Microsystems
Microscopes
The Leica DM IL is a fluorescence ready digital imaging solution designed for use in a variety of laboratory settings including cell culture, micromanipulation, immuno staining and routine live imaging.
  • Inverted laboratory microscope with LED illumination
  • High-performance optics
  • Ergonomic design
  • 3W white light LED illumination
The DM IL LED Inverted Microscope Lab Package is a robust inverted microscope with LED illumination designed for years of use without the need to replace bulbs.
The DM IL LED is fully configured for both brightfield andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable