Quartz Injector, 0.8 mm Int. Ø for Optima 43/53/7300V, PerkinElmer
Supplier: PerkinElmer
N0695461 N0695461
CA97012-888EA 418.59 CAD
97012-888 CA97012-888
Quartz Injector, 0.8 mm Int. Ø for Optima 43/53/7300V, PerkinElmer
Microinjectors Microinjectors, Programmable
This Quartz injector is for use with Optima® 4300V/5300V/7300V ICP-OES series instruments.
Quartz injectors are designed for organic solvents and non-HF matrices.