Viton O-Rings - Size 009, Pkg. 5, PerkinElmer
Supplier: PerkinElmer
L1003006 L1003006
CA97012-732PK 29.63 CAD
CA97012-732 97012-732
Viton O-Rings - Size 009, Pkg. 5, PerkinElmer
Seals Sealing Rings
For use at temperatures up to 225 °C.
It is recommended that these lower O-Rings are replaced during TurboMatrix TD/ATD preventative maintenance. Included in the thermal desorber starter kit (M0413541).