MakeBlock mBot-S Explorer Kits
Supplier: THE DOUGLAS STEWART CO.,INC
P1010045
470324-122EA 140.69 CAD
470324-122
MakeBlock mBot-S Explorer Kits
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Entry-level educational robot for STEAM education and targeting students.
  • Contains many electronic modules
  • Updated version of mBot
  • Comes with an LED matrix display
  • Ages 8+
The MakeBlock mBot-S explorer kit is an entry-level educational robot for STEAM education and suitable for students from ages eight and above. It contains many electronic modules such as a LED matrix, sensors, an IR transmitter, and IR receiver. It has a companion programming software mBlock (for PC/mobile) and supports the Makeblock app for 3D construction.
mBot-S is an updated version of mBot. In addition to the features that mBot provides, mBot-S also comes with an LED matrix display. Students can put the LED display in any position as they please: a front, back, at the top. the LED matrix display brings diversity into mBot-S, allowing students to customize images by programming the RBG lights. Students turn the LED display into a board showing scores, weather forecasts, or emotions.