N,N-Dimethylformamide, anhydrous 99.80%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid dimethylamide, DMFA, Formyldimethylamine, N-Formyldimethylamine, Dimethylformamide, NSC 5356, DMF

Packaged under argon.

047390-5Y 047390-AE 047390-K2 047390-M1
AA47390-5YPK 227.06 CAD
AA47390-5Y AA47390-AE AA47390-K2 AA47390-M1
N,N-Dimethylformamide, anhydrous 99.80%, AcroSeal™
N,N-Dimethylformamide

AcroSeal™ septum bottled solvent


  • Supports optimum performance of your moisture-sensitive work

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C
Melting Pt: -61 °C
Density: 0.944
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (GC) ≥99.75%
Water content (Karl Fischer Titration) ≤50 ppm
Residue on evaporation ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR