Tetrahydrofuran, anhydrous 99,8 Stabilised, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Stabilization: 0.025% BHT
Argon_filled

047382-AE 047382-K2 047382-M1
AA47382-AEEA 106.67 CAD
AA47382-AE AA47382-K2 AA47382-M1
Tetrahydrofuran, anhydrous 99,8 Stabilised, AcroSeal™
Tetrahydrofuran

AcroSeal™ septum bottled solvent.


  • Supports optimum performance of your moisture-sensitive work

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.889
Flash Pt: –21°C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (GC) ≥99.8%
Water content (Karl Fischer Titration) ≤50 ppm
Stabilizer BHT

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR