Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal™ BHT-free, over molecular sieves

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butylene oxide, THF, Diethylene oxide

Packaged under argon.

047327-AE 047327-K2
AA47327-AEEA 107.93 CAD
AA47327-AE AA47327-K2
Tetrahydrofuran, anhydrous ≥99.8%, AcroSeal™ BHT-free, over molecular sieves
Tetrahydrofuran

AcroSeal™ septum bottled solvent


  • Supports optimum performance of your moisture-sensitive work

Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66
Melting Pt: -108.5
Density: 0.889
Flash Pt: –21°C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear Colorless liquid
Assay (GC) ≥99.8%
Water content (Karl Fischer Titration) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR