VWR® Transfer Caps for Shaker Flasks
Supplier: VWR International
76404-502 76404-504 76404-506
76404-502CS 2055.48 CAD
76404-502 76404-504 76404-506
VWR® Transfer Caps for Shaker Flasks
Closures Bottle Closures
These transfer caps are for the VWR® 5 L shaker flasks. Many caps are available for various applications.
  • Inverted transfer caps use gravity to transfer which maintains higher culture viability
  • Bi-directional caps use a standard pump to transfer culture or media
Transfer caps allow quick and sterile cell transfer between flask and vessel.