VWR® Shaker Flasks
Supplier: VWR International
76404-498 76404-500
76404-498CS 845.53 CAD
76404-498 76404-500
VWR® Shaker Flasks
Flasks Erlenmeyer Flasks
VWR® 5 L shaker flasks are perfect for shaker culture applications and storage.
  • Plug seal and vent caps are available
  • Vent caps with 0.2 µm hydrophobic filters for gas exchange without contamination
  • Polycarbonate bottle has a high standard of transparency, clear scale and allows easy observation
  • Graduated markings
  • Packaged individually and sterilized by E-Beam
  • Non-pyrogenic
Ideal for shaker culture applications, the recommended volume is 30 to 40% of the bottle volume.