Laboratory Glassware Washer, PLW6111 SlimLine, Miele
Supplier: Miele
62611177US 62611176US 62611177US 99999529 99999530
76397-798EA 54423.94 CAD
76397-798 76397-800 CA76397-798 CA76397-800 CA77445-688 CA77445-690
Laboratory Glassware Washer, PLW6111 SlimLine, Miele
Glassware Washers
Slimline Laboratory glassware washer with compact dimensions and high capacity.
  • Performance/cycle 126 narrow-neck glasses or 121 pipettes
  • Ergonomic loading and unloading telescopic runners
  • Flexible: Four different level positions
  • Have everything in view thanks to the full-glass door
SlimLine design: The slim design, just 23³/₅" wide, fits in the smallest of spaces while ensuring a high capacity.
Full-glass door: In combination with the chamber lighting, the glass door allows a visual check of the current status.
Conductivity monitoring (76397-798 only): Reliably clean: Residue-free final rinse for perfect results. Residues from chemical agents during the final rinse phase can compromise the reprocessing result. Conductivity monitoring of the wash water facilitates the detection of such residues and their reduction to a defined level through further rinsing. This means that the reprocessing is more reliable, more resource-friendly and also reproducible.
Certifications: EN 61010-1, EN 601010-1, CSA approved, CE-marked.
Ordering information: Includes drain pump, boiler, steam condenser, chamber lighting, Hepa H14, printer, sampling tap and 2 integrated dispensing pumps note only 76397-798 has a conductivity sensor. Baskets and accessories are not included when purchasing just the glassware washer, these must be ordered separately. Please contact your Avantor Sales representative/technical services for assistance with the product configuration. Packages including glassware washer, accessories and chemicals are also available please see details below.