SHIRACLEAN Industrial Ultrasonic Cleaners with Filtration, Elma
Supplier: Elma
TVT-025G TVT-033G TVT-045G TVT-086G TVT-110G
76326-640EA 28552.78 CAD
76326-640 76326-642 76326-644 76326-648 76326-654 76326-656
SHIRACLEAN Industrial Ultrasonic Cleaners with Filtration, Elma
Ultrasonic Cleaners
SHIRACLEAN high capacity industrial ultrasonic cleaners are designed to clean a wide range of parts in industrial shops and laboratories. Application areas include removal of chips and coolant from machined parts, solder flux removal, injection mold cleaning, and 3-D mold support removal.
  • Fully insulated stainless steel tank with Hammertone powder coat finished exterior
  • Stainless steel work area, plumbing, basket and hinged cover
  • Digital temperature control
  • Low liquid level sensor
  • Illuminated control panel
  • Powerful high capacity ultrasonic tank with
More Product Information