Forceps, Gemologist, Standard, Excelta Corp®
Supplier: Excelta
JS-2 JS-3 JS-4 JS-5 JS-7 TRISISTA3 TRISISTA7
75880-478EA 115.64 CAD
75880-478 75880-480 75880-482 75880-484 75880-488 75880-506 75880-508
Forceps, Gemologist, Standard, Excelta Corp®
Forceps
Designed for handling gem stones.
  • Gemologist forceps for fine gemstones
  • Fluted grips for positive control
  • Stainless steel construction
These straight Gemologist forceps are designed to handle extra fine gems. The handles are serrated for better control. These are made of AISI 440 Stainless Steel except for the Trisista, which are made of AISI 316L Stainless Steel, which is anti-magnetic.